Merck
Stabilization of zwitterionic aryltrifluoroborates against hydrolysis.
Chemical communications (Cambridge, England) (2010-08-07)
Casey R Wade, Haiyan Zhao, François P Gabbaï
ABSTRACT
The presence of a proximal cationic group in zwitterionic aryltrifluoroborates such as o-(Ph(2)MeP)C(6)H(4)(BF(3)) stabilizes these compounds against hydrolysis.
Product Description
Supelco
Boron trifluoride - 1-butanol solution, ~10% in 1-butanol (∼1.3 M), LiChropur