Merck
  • Controlled synthesis of luminescent polymers using a bis-dithiobenzoate RAFT agent.
Controlled synthesis of luminescent polymers using a bis-dithiobenzoate RAFT agent.
Chemical communications (Cambridge, England) (2008-02-23)
Ming Chen, Kenneth P Ghiggino, Ezio Rizzardo, San H Thang, Gerard J Wilson
ABSTRACT
A higher efficiency of excitation energy transfer occurs to a luminescent diphenylanthracenyl acceptor incorporated at the centre, rather than the end, of an acenaphthylene polymer chain.
Product Description
Supelco
Acenaphthylene, analytical standard
Sigma-Aldrich
Acenaphthylene, 75%
Sigma-Aldrich
Acenaphthylene, 99%