Merck
(Immuno)histological Analysis of Ewing Sarcoma.
Methods in molecular biology (Clifton, N.J.) (2020-12-17)
David Marcilla, Isidro Machado, Thomas G P Grünewald, Antonio Llombart-Bosch, Enrique de Álava
ABSTRACT
The diagnosis of Ewing sarcoma requires the integration of the information generated from numerous techniques, some of them being very sophisticated. However, the first steps of the diagnostic process are crucial to achieve the maximum possible diagnostic performance. In this chapter we will review how to handle the diagnostic specimen from its collection, how to prepare it for diagnosis, how to make a complete pathology report, and provide guidance for the reasonable use of immunohistochemical techniques in this malignancy.
Sigma-Aldrich
Anti-CCNB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GLG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution