Merck
  • Hard Coating Materials Based on Photo-Reactive Silsesquioxane for Flexible Application: Improvement of Flexible and Hardness Properties by High Molecular Weight.
Hard Coating Materials Based on Photo-Reactive Silsesquioxane for Flexible Application: Improvement of Flexible and Hardness Properties by High Molecular Weight.
Polymers (2021-06-03)
Jong Tae Leem, Woong Cheol Seok, Ji Beom Yoo, Sangkug Lee, Ho Jun Song
ABSTRACT
EPOSS of polyhedral oligomeric silsesquioxanes (POSS) mixture structure and LPSQ of ladder-like polysilsesquioxane (LPSQ) structure were synthesized via sol-gel reaction. EPSQ had a high molecular weight due to polycondensation by potassium carbonate. The EPSQ film showed uniform surface morphology due to regular double-stranded structure. In contrast, the EPOSS-coated film showed nonuniform surface morphology due to strong aggregation. Due to the aggregation, the EPOSS film had shorter d-spacing (d1) than the EPSQ film in XRD analysis. In pencil hardness and nanoindentation analysis, EPSQ film showed higher hardness than the EPOSS film due to regular double-stranded structure. In addition, in the in-folding (r = 0.5 mm) and out-folding (r = 5 mm) tests, the EPSQ film did not crack unlike the EPOSS coated film.
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O