Merck
  • Specific chemotaxis of magnetically labeled mesenchymal stem cells: implications for MRI of glioma.
Specific chemotaxis of magnetically labeled mesenchymal stem cells: implications for MRI of glioma.
Molecular imaging and biology (2012-03-16)
Margaret F Bennewitz, Kevin S Tang, Eleni A Markakis, Erik M Shapiro
ABSTRACT
Glioblastoma multiforme (GBM) is a lethal disease marked by infiltration of cancerous cells into the surrounding normal brain. The dire outcome of GBM patients stems in part from the limitations of current neuroimaging methods. Notably, early cancer detection methodologies are lacking, without the ability to identify aggressive, metastatic tumor cells. We propose a novel approach for tumor detection using magnetic resonance imaging (MRI) based on imaging specific tumor tropism of mesenchymal stem cells (MSCs) labeled with micron-sized iron oxide particles (MPIOs). MPIO labeled and unlabeled MSCs were compared for viability, multi-lineage differentiation, and migration, where both chemotactic and chemokinetic movement were assessed in the presence of serum-free medium, serum-containing medium, and glioma-conditioned medium. MRI was performed on agarose samples, consisting of MPIO-labeled single MSCs, to confirm the capability to detect single cells. We determined that MPIO-labeled MSCs exhibit specific and significant chemotactic migration towards glioma-conditioned medium in vitro. Confocal fluorescence microscopy confirmed that MPIOs are internalized and do not impact important cell processes of MSCs. Lastly, MPIO-labeled MSCs appear as single distinct, dark spots on T(2)*-weighted MRI, supporting the robustness of this contrast agent for cell tracking. This is the first study to show that MPIO-labeled MSCs exhibit specific tropism toward tumor-secreted factors in vitro. The potential for detecting single MPIO-labeled MSCs provides rationale for in vivo extension of this methodology to visualize GBM in animal models.
Sigma-Aldrich
Rat Mesenchymal Stem Cells (Bone Marrow), These primary bone marrow derived Rat Mesenchymal Stem Cells isolated from the iliac crest of adult Fisher 344 rats provide a convenient source of multipotent rat mesenchymal stem cells.
Pricing and availability is not currently available.