Merck
  • The antimalarial MMV688533 provides potential for single-dose cures with a high barrier to Plasmodium falciparum parasite resistance.
The antimalarial MMV688533 provides potential for single-dose cures with a high barrier to Plasmodium falciparum parasite resistance.
Science translational medicine (2021-07-23)
James M Murithi, Cécile Pascal, Jade Bath, Xavier Boulenc, Nina F Gnädig, Charisse Flerida A Pasaje, Kelly Rubiano, Tomas Yeo, Sachel Mok, Sylvie Klieber, Paul Desert, María Belén Jiménez-Díaz, Jutta Marfurt, Mélanie Rouillier, Mohammed H Cherkaoui-Rbati, Nathalie Gobeau, Sergio Wittlin, Anne-Catrin Uhlemann, Ric N Price, Grennady Wirjanata, Rintis Noviyanti, Patrick Tumwebaze, Roland A Cooper, Philip J Rosenthal, Laura M Sanz, Francisco Javier Gamo, Jayan Joseph, Shivendra Singh, Sridevi Bashyam, Jean Michel Augereau, Elie Giraud, Tanguy Bozec, Thierry Vermat, Gilles Tuffal, Jean-Michel Guillon, Jérôme Menegotto, Laurent Sallé, Guillaume Louit, Marie-José Cabanis, Marie Françoise Nicolas, Michel Doubovetzky, Rita Merino, Nadir Bessila, Iñigo Angulo-Barturen, Delphine Baud, Lidiya Bebrevska, Fanny Escudié, Jacquin C Niles, Benjamin Blasco, Simon Campbell, Gilles Courtemanche, Laurent Fraisse, Alain Pellet, David A Fidock, Didier Leroy
ABSTRACT
The emergence and spread of Plasmodium falciparum resistance to first-line antimalarials creates an imperative to identify and develop potent preclinical candidates with distinct modes of action. Here, we report the identification of MMV688533, an acylguanidine that was developed following a whole-cell screen with compounds known to hit high-value targets in human cells. MMV688533 displays fast parasite clearance in vitro and is not cross-resistant with known antimalarials. In a P. falciparum NSG mouse model, MMV688533 displays a long-lasting pharmacokinetic profile and excellent safety. Selection studies reveal a low propensity for resistance, with modest loss of potency mediated by point mutations in PfACG1 and PfEHD. These proteins are implicated in intracellular trafficking, lipid utilization, and endocytosis, suggesting interference with these pathways as a potential mode of action. This preclinical candidate may offer the potential for a single low-dose cure for malaria.
Product Description
Supelco
Anhydrotetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture