Merck
  • SIRT5 impairs aggregation and activation of the signaling adaptor MAVS through catalyzing lysine desuccinylation.
SIRT5 impairs aggregation and activation of the signaling adaptor MAVS through catalyzing lysine desuccinylation.
The EMBO journal (2020-04-18)
Xing Liu, Chunchun Zhu, Huangyuan Zha, Jinhua Tang, Fangjing Rong, Xiaoyun Chen, Sijia Fan, Chenxi Xu, Juan Du, Junji Zhu, Jing Wang, Gang Ouyang, Guangqing Yu, Xiaolian Cai, Zhu Chen, Wuhan Xiao
ABSTRACT
RLR-mediated type I IFN production plays a pivotal role in innate antiviral immune responses, where the signaling adaptor MAVS is a critical determinant. Here, we show that MAVS is a physiological substrate of SIRT5. Moreover, MAVS is succinylated upon viral challenge, and SIRT5 catalyzes desuccinylation of MAVS. Mass spectrometric analysis indicated that Lysine 7 of MAVS is succinylated. SIRT5-catalyzed desuccinylation of MAVS at Lysine 7 diminishes the formation of MAVS aggregation after viral infection, resulting in the inhibition of MAVS activation and leading to the impairment of type I IFN production and antiviral gene expression. However, the enzyme-deficient mutant of SIRT5 (SIRT5-H158Y) loses its suppressive role on MAVS activation. Furthermore, we show that Sirt5-deficient mice are resistant to viral infection. Our study reveals the critical role of SIRT5 in limiting RLR signaling through desuccinylating MAVS.
Sigma-Aldrich
Succinyl coenzyme A sodium salt, ≥85%