Merck
  • Synthesis, in vitro, and in silico evaluation of organometallic technetium and rhenium thymidine complexes with retained substrate activity toward human thymidine kinase type 1.
Synthesis, in vitro, and in silico evaluation of organometallic technetium and rhenium thymidine complexes with retained substrate activity toward human thymidine kinase type 1.
Journal of medicinal chemistry (2008-10-08)
Dominique Desbouis, Harriet Struthers, Vojtech Spiwok, Tatiana Küster, Roger Schibli
ABSTRACT
Human cytosolic thymidine kinase (hTK1) has proven to be a suitable target for noninvasive imaging of cancer cell proliferation using radiolabeled substrates such as [ (18)F]fluorothymidine ([ (18)F]FLT). However, a thymidine tracer useful for single photon emission tomography (SPECT) based on the inexpensive radionuclide technetium-99m would be of significant interest. In this work, a series of thymidine derivatives labeled with the organometallic [M(CO) 3] (+) core (M = (99m)Tc, Re) were synthesized. Neutral, cationic, and anionic complexes were readily formed in aqueous media, and all were substrates of recombinant hTK1 when incubated with ATP. The neutral complexes were phosphorylated to a greater extent than the charged complexes. The extent of phosphorylation was further improved by increasing the spacer length separating thymidine and the organometallic core. A molecular dynamics simulation study performed with a modified hTK1 structure supported the experimental findings. In vitro cell internalization experiments performed in a human neuroblastoma cell line (SKNMC) showed low uptake of the charged complexes but significant uptake for the neutral, lipophilic complexes with a log P value >1.
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture