Merck
  • Prolonged activation of NF-kappaB by human cytomegalovirus promotes efficient viral replication and late gene expression.
Prolonged activation of NF-kappaB by human cytomegalovirus promotes efficient viral replication and late gene expression.
Virology (2005-11-24)
Ian B DeMeritt, Jagat P Podduturi, A Michael Tilley, Maciej T Nogalski, Andrew D Yurochko
ABSTRACT
Infection of fibroblasts by human cytomegalovirus (HCMV) rapidly activates the NF-kappaB signaling pathway, which we documented promotes efficient transactivation of the major immediate-early promoter (DeMeritt, I.B., Milford, L.E., Yurochko, A.D. (2004). Activation of the NF-kappaB pathway in human cytomegalovirus-infected cells is necessary for efficient transactivation of the major immediate-early promoter. J. Virol. 78, 4498-4507). Because a second, sustained increase in NF-kappaB activity following the initial phase of NF-kappaB activation was also observed, we investigated the role that this prolonged NF-kappaB activation played in viral replication and late gene expression. We first investigated HCMV replication in cells in which NF-kappaB activation was blocked by pretreatment with NF-kappaB inhibitors: HCMV replication was significantly decreased in these cultures. A decrease in replication was also observed when NF-kappaB was inhibited up to 48 h post-infection, suggesting a previously unidentified role for NF-kappaB in the regulation of the later class of viral genes.
Sigma-Aldrich
Lactacystin, Synthetic, Lactacystin, Synthetic, CAS 1258004-00-0, is an irreversible inhibitor of 20S proteasome inhibitor (IC₅₀ = 500 nM). A covalent inhibitor of the chymotrypsin & trypsin-like activities of proteasome.
Sigma-Aldrich
Anti-BrdU (Ab-3) Mouse mAb (Mobu-1), liquid, clone Mobu-1, Calbiochem®
Sigma-Aldrich
BAY 11-7082, The BAY 11-7082, also referenced under CAS 19542-67-7, controls the biological activity of TNF-α. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Sigma-Aldrich
MG-132(R), ≥95% (HPLC)