Merck
  • Nucleus Accumbens Tac1-Expressing Neurons Mediate Stress-Induced Anhedonia-like Behavior in Mice.
Nucleus Accumbens Tac1-Expressing Neurons Mediate Stress-Induced Anhedonia-like Behavior in Mice.
Cell reports (2020-11-05)
Zi-Xuan He, Yue-Yue Yin, Ke Xi, Zhen-Kai Xing, Jian-Bo Cao, Ting-Yu Liu, Lin Liu, Xiao-Xiao He, Hua-Li Yu, Xiao-Juan Zhu
ABSTRACT
Major depressive disorder (MDD) presents with two primary symptoms: depressed mood and anhedonia, which suggests that distinct neuronal circuits may regulate MDD. However, the underlying circuits of these individual symptoms linked to depression remain elusive. Herein, we identify a discrete circuit of tachykinin precursor 1 (Tac1)-expressing neurons in the nucleus accumbens (NAc) lateral shell, which project to ventral pallidum and contribute to stress-induced anhedonia-like behavior. Selective inhibition and activation of Tac1NAc neurons bidirectionally modulate stress susceptibility, revealing that Tac1 neurons in the NAc are critical for regulating anhedonia-like behaviors. We find that a subpopulation of VP neurons receives inhibitory inputs from Tac1NAc neurons and exhibits decreased excitability in susceptible mice. Furthermore, the inhibition of the neurokinin 1 receptor promotes susceptibility to social stress. Overall, our study reveals a discrete circuit regulating anhedonia-like behavior in mice.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Synapsin I Antibody, Chemicon®, from rabbit