Merck
  • High density cultivation for efficient sesquiterpenoid biosynthesis in Synechocystis sp. PCC 6803.
High density cultivation for efficient sesquiterpenoid biosynthesis in Synechocystis sp. PCC 6803.
Scientific reports (2020-04-05)
Dennis Dienst, Julian Wichmann, Oliver Mantovani, João S Rodrigues, Pia Lindberg
ABSTRACT
Cyanobacteria and microalgae are attractive photoautotrophic host systems for climate-friendly production of fuels and other value-added biochemicals. However, for economic applications further development and implementation of efficient and sustainable cultivation strategies are essential. Here, we present a comparative study on cyanobacterial sesquiterpenoid biosynthesis in Synechocystis sp. PCC 6803 using a commercial lab-scale High Density Cultivation (HDC) platform in the presence of dodecane as in-situ extractant. Operating in a two-step semi-batch mode over a period of eight days, volumetric yields of (E)-α-bisabolene were more than two orders of magnitude higher than previously reported for cyanobacteria, with final titers of 179.4 ± 20.7 mg * L-1. Likewise, yields of the sesquiterpene alcohols (-)-patchoulol and (-)-α-bisabolol were many times higher than under reference conditions, with final titers of 17.3 ± 1.85 mg * L-1 and 96.3 ± 2.2 mg * L-1, respectively. While specific productivity was compromised particularly for (E)-α-bisabolene in the HDC system during phases of high biomass accumulation rates, volumetric productivity enhancements during linear growth at high densities were more pronounced for (E)-α-bisabolene than for the hydroxylated terpenoids. Together, this study provides additional insights into cell density-related process characteristics, introducing HDC as highly efficient strategy for phototrophic terpenoid production in cyanobacteria.
Product Description
Supelco
(−)-α-Bisabolol, analytical standard
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)