Merck
  • Activation of macrophages by gliadin fragments: isolation and characterization of active peptide.
Activation of macrophages by gliadin fragments: isolation and characterization of active peptide.
Journal of leukocyte biology (2002-04-03)
Ludmila Tucková, Jana Novotná, Petr Novák, Zuzana Flegelová, Tomás Kveton, Lenka Jelínková, Zdenek Zídek, Petr Man, Helena Tlaskalová-Hogenová
ABSTRACT
Celiac disease, induced by dietary gluten, is characterized by mucosal atrophy and local inflammation associated with cell infiltration and activation. Unlike other food proteins, gluten and its proteolytic fragments, besides inducing a specific immune response, were shown to activate components of innate immunity and cause, e.g., direct stimulation of TNF-alpha and IL-10 and a significant rise in NO production by peritoneal macrophages. The identity of the active fragments was established by separating the peptic digest of gliadin by RP-HPLC chromatography. The purest fraction with the highest activity was analyzed by mass spectrometry, and the gliadin peptide sequence was identified as VSFQQPQQQYPSSQ. This peptide (T) and its N- and C-terminally shortened forms (A, B, C and D, E, F) were synthesized. Peptide B (FQQPQQQYPSSQ) elicited the highest TNF-alpha, IL-10, and RANTES secretion and increase in IFN-gamma-primed NO production by mouse macrophages. In contrast, C-terminally shortened peptides had a lower ability to stimulate macrophages than the native form.
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid
Pricing and availability is not currently available.