Merck
  • Protective effect of C4a against hyperoxic lung injury via a macrophage-dependent but not a neutrophil/lymphocyte-dependent signaling pathway.
Protective effect of C4a against hyperoxic lung injury via a macrophage-dependent but not a neutrophil/lymphocyte-dependent signaling pathway.
Molecular medicine reports (2015-12-10)
Xiaoming Jiang, Yubo Ma, Jinfeng Yu, Haihong Li, Fengjie Xie
ABSTRACT
Complement anaphylatoxins have been investigated extensively; however, the role of complement anaphylatoxin C4a in hyperoxic lung injury has yet to be investigated. To the best of our knowledge, the present study is the first to demonstrate the role of C4a in hyperoxic lung injury in vitro and in vivo. BALB/c mice were ventilated with 100% oxygen with or without C4a treatment for 36 h. The body weight and the relative lung weight of the mice were determined, along with any morphological changes in the lung. The expression levels of interleukin (IL)-1, IL-6 and tumor necrosis factor-α (TNF-α) were quantified in the lung tissue and bronchoalveolar lavage fluid (BALF) samples by enzyme-linked immunosorbent assay (ELISA) and western blot analysis. The total cell count and the number of macrophages, neutrophils and lymphocytes in the BALF were determined using cytocentrifuge slides and a hemocytometer. Histamine release from total cells in the BALF was also analyzed. The relative mRNA expression levels of CD68, F4/80, CD64, CD19 and CD3 in the murine lung tissue were assessed by reverse transcription-quantitative polymerase chain reaction. The results revealed that hyperoxia induced lung injury and morphological changes, and increased the expression levels of IL-1, IL-6 and TNF-α, histamine release, the number of inflammatory cells, and the expression levels of CD68, F4/80, CD64, CD19 and CD3. The hyperoxia-induced morphological changes and inflammatory reaction were significantly attenuated in mice treated with C4a. Treatment with C4a also attenuated the increase in the total cell count, decreased the number of macrophages in the BALF, and suppressed the elevated mRNA expression levels of CD68 and F4/80 in the lung tissue samples. Conversely, treatment with C4a did not affect the number of neutrophils or lymphocytes in the BALF or the mRNA expression of CD64, CD19 and CD3 in lung tissue. In conclusion, C4a attenuated hyperoxic lung injury via a macrophage-dependent but not a neutrophil/lymphocyte-dependent pathway.
Sigma-Aldrich
Anti-Tumor Necrosis Factor-α antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Interleukin-6 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution