Merck
  • Trichostatin A induces 5-lipoxygenase promoter activity and mRNA expression via inhibition of histone deacetylase 2 and 3.
Trichostatin A induces 5-lipoxygenase promoter activity and mRNA expression via inhibition of histone deacetylase 2 and 3.
Journal of cellular and molecular medicine (2011-09-03)
Laura Pufahl, Careen Katryniok, Nicole Schnur, Bernd L Sorg, Julia Metzner, Manuel Grez, Dieter Steinhilber
ABSTRACT
The 5-lipoxygenase (5-LO) is the key enzyme in the formation of leukotrienes. We have previously shown that the histone deacetylase (HDAC) inhibitor trichostatin A (TSA) activates 5-LO transcription via recruitment of Sp1, Sp3 and RNA polymerase II to the proximal promoter. To identify the HDACs involved in the regulation of 5-LO promoter activity isoform-specific HDAC inhibitors were applied. 5-LO promoter activity and mRNA expression were up-regulated by the class I HDAC inhibitors apicidin and MS-275 but not by class II inhibitors. Knockdown of HDAC 1, 2 and 3 revealed that HDAC2 and HDAC3 but not HDAC1 is involved in the up-regulation of 5-LO mRNA expression. To analyse the chromatin modifications at the 5-LO promoter associated with HDAC inhibition, the time course of 5-LO mRNA induction by trichostatin A was investigated and the concomitant changes in histone modifications at the 5-LO promoter in HL-60, U937 and Mono Mac6 cells were determined. Chromatin immunoprecipitation analysis revealed that trichostatin A increases acetylation of histones H3 and H4 at the 5-LO core promoter in HL-60 and U937 cells whereas no significant changes were observed in Mono Mac6 cells. The appearance of H3 and H4 acetylation preceded the 5-LO mRNA induction whereas in all three cell lines, induction of 5-LO mRNA expression correlated with histone H3 lysine 4 trimethylation (H3K4me3), a marker for transcriptional activity of gene promoters.
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Apicidin, ≥98% (HPLC), from microbial