Merck
  • Epitopes recognized by antibodies to denatured core protein of hepatitis B virus.
Epitopes recognized by antibodies to denatured core protein of hepatitis B virus.
Molecular immunology (1993-02-01)
V Bichko, F Schödel, M Nassal, E Gren, I Berzinsh, G Borisova, S Miska, D L Peterson, E Gren, P Pushko
ABSTRACT
Particulate and denatured core protein as well as e-antigen (HBe) of hepatitis B virus (HBV) differ in part immunologically but this has not been studied in sufficient detail. Therefore, in this study the B-cell immune response to native and denatured HBV core protein which both can exhibit HBe-specific epitopes was examined using a panel of mouse MABs and rabbit polyclonal antibodies to native and denatured core protein and polyclonal anti-HBe/anti-HBc antibodies from sera of infected patients. Epitope mapping was performed using a set of partially overlapping synthetic HBc peptides, carboxy-terminally truncated HBc proteins and various HBc fusion proteins. A major immunogenic region between amino acids 134-140 and two less immunogenic regions, one spanning amino acids 2-10 and one with three partially overlapping epitopes between amino acid positions 138 and 154, were defined by mouse MABs. Polyclonal rabbit antibodies to denatured HBc, woodchuck and ground squirrel hepatitis core proteins (WHc and GSHc) recognized similar epitopes but in addition occasionally region 61-85, and the latter was also recognized on particulate HBc. Two antigenic regions (amino acid positions 2-10 and 138-145) were found to be exposed on HBe from human serum, and were recognized by mouse anti-HBe but not by anti-HBc antibodies from sera of infected patients. This study demonstrates a more complex pattern of HBc and HBe epitopes than detected previously and provides tools to study conformational changes which may take place during HBc/HBe processing, transport and core particle assembly.
Sigma-Aldrich
Anti-Hepatitis B Virus Antibody, core Antigen (ayw), clone 10E11, a.a. 2-10, ascites fluid, clone 10E11, Chemicon®
Sigma-Aldrich
Anti-Hepatitis B Virus Antibody, core Antigen (ayw), clone 14E11, a.a. 134-140, ascites fluid, clone 14E11, Chemicon®