Merck
  • Development and characterization of a camelid single-domain antibody directed to human CD22 biomarker.
Development and characterization of a camelid single-domain antibody directed to human CD22 biomarker.
Biotechnology and applied biochemistry (2018-03-16)
Fatemeh Faraji, Nader Tajik, Mahdi Behdani, Mohammad Ali Shokrgozar, Amir Hassan Zarnani, Fatemeh Shahhosseini, Mahdi Habibi-Anbouhi
ABSTRACT
CD22 is a B-cell-specific trans-membrane glycoprotein, which is found on the surface of the most B cells and modulates their function, survival, and apoptosis. Recently, targeting this cell surface biomarker in B-cell malignancies and disorders has attracted a lot of attention. The variable domain of camelid single-chain antibodies (VHH, nanobody) is a form of antibodies with novel properties including small size (15-17 kDa), thermal and chemical stability, high affinity and homology to human antibody sequences. In this study, a novel anti-CD22-specific VHH (Nb) has been developed and characterized by the screening of an immunized phage display library and its binding to CD22+ B cells is evaluated. Produced anti-CD22 VHH had a single protein band about 17 kDa of molecular size in Western blotting and its binding affinity was approximately 9 × 10-9  M. Also, this product had high specificity and it was able to recognize the natural CD22 antigen in CD22+ cell lysate as well as on the cell surface (93%). This anti-CD22 VHH with both high affinity and specificity recognizes CD22 antigen well and can be used in diagnosis and treatment of B cell disorders and malignancies.
Sigma-Aldrich
CD47 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)