
Trimethoprim Related Compound A

Synonym(s):
4-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-2-ol
Empirical Formula (Hill Notation):
C14H17N3O4
CAS Number:
Molecular Weight:
291.30
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2