
trans-2-Bromovinylboronic acid MIDA ester

Synonym(s):
BB1
Empirical Formula (Hill Notation):
C7H9BBrNO4
CAS Number:
Molecular Weight:
261.87
Products
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing