
Tizanidine hydrochloride

Synonym(s):
5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine hydrochloride
Empirical Formula (Hill Notation):
C9H9Cl2N5S
CAS Number:
Molecular Weight:
290.17
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3