
Sulfamethoxypyridazine

Synonym(s):
N1-(6-Methoxypyridazin-3-yl)sulfanilamide
Empirical Formula (Hill Notation):
C11H12N4O3S
CAS Number:
Molecular Weight:
280.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing