
Santalol, α- and β-isomers

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing