
Ranitidine Related Compound C

Synonym(s):
Ranitidine S-oxide, N-{2-{{{5-[(Dimethylamino)methyl]furan-2-yl}methyl}sulfinyl}ethyl}-N′-methyl-2-nitroethene-1,1-diamine
Empirical Formula (Hill Notation):
C13H22N4O4S
CAS Number:
Molecular Weight:
330.40
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2