
Nicergoline for peak identification

Synonym(s):
Nicergoline, 5-Bromonicotinic acid 10-methoxy-1,6-dimethylergoline-8-methyl ester
Empirical Formula (Hill Notation):
C24H26BrN3O3
CAS Number:
Molecular Weight:
484.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing