
Nevirapine anhydrous

Synonym(s):
Nevirapine, 11-Cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2′,3′-e][1,4]diazepin-6-one
Empirical Formula (Hill Notation):
C15H14N4O
CAS Number:
Molecular Weight:
266.30
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing