
Molsidomine impurity B

Synonym(s):
Nitrosomorpholine, NMOR
Empirical Formula (Hill Notation):
C4H8N2O2
CAS Number:
Molecular Weight:
116.12
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing