
Metoprolol Related Compound A

Synonym(s):
(±)-1-(Ethylamino)-3-[4-(2-methoxyethyl)phenoxy]- 2-propanol
Empirical Formula (Hill Notation):
C14H23NO3
CAS Number:
Molecular Weight:
253.34
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2