
Metolachlor, predominantly S-Metolachlor

Synonym(s):
2-Chloro-N-(2-ethyl-6-methylphenyl)-N-[(1S)-2-methoxy-1-methylethyl]acetamide
Empirical Formula (Hill Notation):
C15H22ClNO2
CAS Number:
Molecular Weight:
283.79
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique