
Ipratropium bromide

Synonym(s):
Ipratropium bromide monohydrate, Atropine isopropyl bromide
Empirical Formula (Hill Notation):
C20H30BrNO3 · H2O
CAS Number:
Molecular Weight:
430.38
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3