
Ginkgolide A

Empirical Formula (Hill Notation):
C20H24O9
CAS Number:
Molecular Weight:
408.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing