
Exo-tetrahydrocannabinol

Synonym(s):
exo-THC solution
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing