
Docusate sodium salt

Synonym(s):
AOT, Bis(2-ethylhexyl) sulfosuccinate sodium salt, Sodium bis(2-ethylhexyl) sulfosuccinate, Sulfobutanedioic acid bis(2-ethylhexyl ester) sodium salt, Sulfosuccinic acid bis(2-ethylhexyl) ester sodium salt, ‘Dioctyl’ sulfosuccinate sodium salt
Empirical Formula (Hill Notation):
C20H37NaO7S
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing