
Desogestrel Related Compound A

Synonym(s):
13-Ethyl-11-methylene-18, 19-dinor-5α, 17α-pregn-3-en-20-yn-17-ol
Empirical Formula (Hill Notation):
C22H30O
CAS Number:
Molecular Weight:
310.47
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing