
Dacarbazine Related Compound A

Synonym(s):
5-Amino-4-imidazolecarboxamide hydrochloride, 4-Amino-5-imidazolecarboxamide hydrochloride, AICA
Empirical Formula (Hill Notation):
C4H6N4O · HCl
CAS Number:
Molecular Weight:
162.58
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing