
Dacarbazine impurity B

Synonym(s):
5-Amino-4-imidazolecarboxamide
Empirical Formula (Hill Notation):
C4H6N4O
CAS Number:
Molecular Weight:
126.12
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing