
Cepharanthine

Synonym(s):
(+)-Cepharanthine, 6′,12′-Dimethoxy-2,2′-dimethyl-6,7-(methylenebis(oxy))oxyacanthan, O-Methylcepharanoline, CEP, Cepharanthin, Cepharantin, NSC 623442, [14aS-(14aR*,26aS*)]-2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl- H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2′,3′:17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline
Empirical Formula (Hill Notation):
C37H38N2O6
CAS Number:
Molecular Weight:
606.71
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing