
Caprylic acid

Synonym(s):
Octanoic acid, Acid C8, Caprylic acid
Linear Formula:
CH3(CH2)6COOH
CAS Number:
Molecular Weight:
144.21
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing