
Capecitabine Related Compound A

Synonym(s):
5′-Deoxy-5-fluorocytidine, 4-Amino-1-(5-deoxy-β-D-ribofuranosyl)-5-fluoropyrimidin-2(1H)-one
Empirical Formula (Hill Notation):
C9H12FN3O4
CAS Number:
Molecular Weight:
245.21
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2