
Capecitabine impurity A

Synonym(s):
5′-Deoxy-5-fluorocytidine, 4-Amino-1-(5-deoxy-β-D-ribofuranosyl)-5-fluoropyrimidin-2(1H)-one
Empirical Formula (Hill Notation):
C9H12FN3O4
CAS Number:
Molecular Weight:
245.21
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing