
Boc-Glu-OtBu

Synonym(s):
Boc-L-glutamic acid 1-tert-butyl ester, Boc-Glu-OtBu
Empirical Formula (Hill Notation):
C14H25NO6
CAS Number:
Molecular Weight:
303.35
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing