Bifenazate
Synonym(s):
N′-(4-Methoxybiphenyl-3-yl)hydrazinecarboxylic acid isopropyl ester
Empirical Formula (Hill Notation):
C17H20N2O3
CAS Number:
Molecular Weight:
300.35
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2