Skip to Content
Merck
vibrant-m

Anti-V5 Agarose Affinity Gel antibody produced in mouse

vibrant-m
Synonym(s):
Monoclonal Anti-V5

Products

Product Category
Brand
Biological Source
Clonality
Analyte Suitability
Antibody Form
Conjugate
Quality Segment
Technique
Showing 1-1 of 1