
Amylmetacresol for peak identification

Synonym(s):
Amylmetacresol, 5-Methyl-2-pentylphenol
Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing