
Adapalene Related Compound B

Synonym(s):
Methyl 6-[3-(1-adamantyl)-4-methoxyphenyl]-2-naphthoate
Empirical Formula (Hill Notation):
C29H30O3
CAS Number:
Molecular Weight:
426.55
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2