
Adapalene impurity C

Synonym(s):
o-Adamantylanisole, 1-(2-Methoxyphenyl)-tricyclo[3.3.1.13,7]decane, 2-(Adamant-1-yl)methoxybenzene
Empirical Formula (Hill Notation):
C17H22O
CAS Number:
Molecular Weight:
242.36
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing