
Acitretin Related Compound A

Synonym(s):
(2Z,4E,6E,8E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid, 13-cis-Acitretin, Isoacitretin
Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing