
5,10,15,20-Tetraphenyl-21H, 23H-porphine-p,p′,p,p′′′-tetrasulfonic acid tetrasodium hydrate

Synonym(s):
4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzenesulfonic acid) tetrasodium salt hydrate, meso-Tetraphenylporphine-4,4′,4″,4″′-tetrasulfonic acid tetrasodium salt
Empirical Formula (Hill Notation):
C44H26N4Na4O12S4 · xH2O
CAS Number:
Molecular Weight:
1022.92 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing