Skip to Content
Merck
4-(Trifluoromethyl)styrene 98%, contains 0.1% 4-tert-butylcatechol as inhibitor

4-(Trifluoromethyl)styrene

4-(Trifluoromethyl)styrene 98%, contains 0.1% 4-tert-butylcatechol as inhibitor
Synonym(s):
4-Vinylbenzotrifluoride
Linear Formula:
CF3C6H4CH=CH2
CAS Number:
Molecular Weight:
172.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1