Skip to Content
Merck
3′-Sialyllactose from bovine milk or colostrum, ≥97% (HPLC)

3′-Sialyllactose

3′-Sialyllactose from bovine milk or colostrum, ≥97% (HPLC)
Synonym(s):
α-NeuNAc-(2→3)-β-D-Gal-(1→4)-D-Glc, 3′-N-Acetylneuraminyl-D-lactose sodium salt, 3′-SL, 3′-Sialyl-D-lactose, NANA-Lactose
Empirical Formula (Hill Notation):
C23H39NO19
CAS Number:
Molecular Weight:
633.55

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Tissue
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing