
2,5-Diisopropylphenol

Synonym(s):
2,5-Bis(1-methylethyl)phenol, 2,5-Di(propan-2-yl)phenol
Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing