Skip to Content
Merck
1H,1H,2H,2H-Perfluorodecyltriethoxysilane 97%

1H,1H,2H,2H-Perfluorodecyltriethoxysilane

1H,1H,2H,2H-Perfluorodecyltriethoxysilane 97%
Synonym(s):
Triethoxy-1H,1H,2H,2H-perfluorodecylsilane
Empirical Formula (Hill Notation):
C16H19F17O3Si
CAS Number:
Molecular Weight:
610.38

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing